Thermo Scientific Chemicals

4-Metilbencilfosfonato de dietilo, +98 %, Thermo Scientific Chemicals

Número de catálogo: A15190.09
10 g, Each
Thermo Scientific Chemicals

4-Metilbencilfosfonato de dietilo, +98 %, Thermo Scientific Chemicals

Número de catálogo: A15190.09
10 g, Each
Cantidad
Número de catálogo: A15190.09
también denominado A15190-09
Precio (USD)

Identificadores químicos

CAS
3762-25-2
IUPAC Name
diethyl [(4-methylphenyl)methyl]phosphonate
Molecular Formula
C12H19O3P
InChI Key
QKGBKPZAXXBLJE-UHFFFAOYSA-N
SMILES
CCOP(=O)(CC1=CC=C(C)C=C1)OCC
Form
Liquid
Refractive Index
1.4945-1.4985 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥98.0%

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text