Thermo Scientific Chemicals

Monohidrato de ácido 8-etoxiquinolina-5-sulfónico, 99 %, Thermo Scientific Chemicals

Número de catálogo: A18622.18
50 g, Each
Thermo Scientific Chemicals

Monohidrato de ácido 8-etoxiquinolina-5-sulfónico, 99 %, Thermo Scientific Chemicals

Número de catálogo: A18622.18
50 g, Each
Cantidad
Número de catálogo: A18622.18
también denominado A18622-18
Precio (USD)

Identificadores químicos

CAS
15301-40-3
IUPAC Name
8-ethoxyquinoline-5-sulfonic acid
Molecular Formula
C11H11NO4S
InChI Key
YAMVZYRZAMBCED-UHFFFAOYSA-N
SMILES
CCOC1=C2N=CC=CC2=C(C=C1)S(O)(=O)=O
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale cream to cream to yellow
Water Content (Karl Fischer Titration)
3.43-9.64% (0.5-1.5 waters)
Identification (FTIR)
Conforms

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text