SureClean™ DNA Removal Kit
SureClean™ DNA Removal Kit
SureClean™ DNA Removal Kit
SureClean™ DNA Removal Kit
Reduce free extracellular DNA in food and environmental samples with the SureClean™ DNA Removal Kit. Designed for seamless integration with SureTect™ PCR assays to help minimize false positives and improve workflow efficiency.
Have Questions?
Número de catálogo A65860
Precio (USD)
-
The SureClean™ DNA Removal Kit is an innovative sample pretreatment solution developed for laboratories performing microbiological analysis of food and environmental samples. It specifically targets matrices that experience recurring unconfirmable PCR-positive results. As part of the SureTect™ product portfolio, the kit efficiently reduces free extracellular DNA prior to sample lysis and PCR amplification, helping decrease false-positive results caused by residual DNA contamination.
Key Benefits and Product Features
  • Targets matrices that experience recurring unconfirmable PCR-positive results such as heat-treated, dried, or heavily sanitized samples—particularly PIF, infant nutrition, dairy powders, seafood, ready-to-eat (RTE) meals, dry ingredients, and associated environmental samples
  • Reduces free extracellular DNA in enriched food and environmental samples prior to PCR analysis
  • Helps minimize false-positive PCR results caused by residual free DNA
  • Integrates seamlessly into the SureTect™ molecular testing workflow
  • Supports faster food batch release by reducing unnecessary confirmatory testing
  • Simple 15-minute pretreatment protocol using the Applied Biosystems™ SimpliAmp™ Thermal Cycler
  • Compatible with SureTect™ PCR assays for:
    • Salmonella species—AFNOR: Multi-component foods and specific ingredients, powdered infant formula, infant cereals and related ingredients, and environmental samples; AOAC: Egg products (including egg powder), powdered infant formula, and environmental samples
    • Listeria species—AFNOR: Powdered infant formula, infant cereals and related ingredients, dairy products, and environmental samples; AOAC: Powdered infant formula and environmental samples
    • Listeria monocytogenes—AFNOR: Powdered infant formula, infant cereals and related ingredients, dairy products, and environmental samples; AOAC: Powdered infant formula and environmental samples
    • Cronobacter species—AFNOR: Powdered infant formula, infant cereals, related ingredients, and environmental samples; AOAC: Powdered infant formula and environmental samples
    • Staphylococcus aureus—AOAC: Powdered infant formula, whey protein concentrate, and whole milk powder
    • Vibrio cholerae, V. parahaiolyticus, and V. vulnificus—AOAC: Cooked shrimp and raw mussels
  • Includes sufficient reagents for 96 DNA removal treatments
  • Designed for use with the Applied Biosystems™ SimpliAmp™ Thermal Cycler and QuantStudio™ 5 Food Safety Real-Time PCR System with RapidFinder™ Analysis Software v3.1 or later
  • Optimized for laboratories performing microbiological testing of food and environmental samples
Únicamente para las pruebas de muestras alimentarias y ambientales.
Especificaciones
Para utilizar conSureTect™ Cronobacter species PCR Assay (Cat. No. A56845), SureTect™ Listeria monocytogenes PCR Assay (Cat. No. A56843), SureTect™ Listeria species PCR Assay (Cat. No. A56842), SureTect™ Salmonella species PCR Assay (Cat. No. A56841), SureTect™ Staphylococcus aureus PCR Assay (Cat. No. A56839), SureTect™ Vibrio cholerae, V. parahaemolyticus and V. vulnificus PCR Assay (Cat. No. A56837)
Para utilizar con (equipo)Applied Biosystems™ SimpliAmp™ Thermal Cycler (Cat. No. A55333) and Applied Biosystems™ QuantStudio™ 5 Food Safety Real-Time PCR System (Cat. No. A36320; A36328) with Thermo Scientific™ RapidFinder™ Analysis Software v3.1 or later (Cat. No. A53289)
FormatoKit, SureClean™ Tubes with SureClean™ solution, 96-well Pierceable Seals
Compatibilidad de alto rendimientoHigh-throughput Compatible
Nº de pruebas96 Tests
Línea de productosSureClean™ and SureTect™
Tipo de productoDNA Removal Kit
Categoría de investigaciónFood Microbiology
Para utilizar con (aplicación)Real Time PCR (qPCR)
Unit SizeEach
Contenido y almacenamiento
Store the SureClean™ DNA Removal Kit at -20°C in a non-frost-free freezer for long-term storage.