Bacto™ CD Supreme Feed (2X), liquid
Gibco™

Bacto™ CD Supreme Feed (2X), liquid

Chemically defined nutrient feed for optimized E. coli plasmid DNA and protein production.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
A937821010 x 500 mL
A9378201500 mL
Número de catálogo A9378210
Precio (USD)
-
Cantidad:
10 x 500 mL

Gibco Bacto CD Supreme Feed (2X) is an animal origin–free, chemically defined microbial feed specifically formulated to extend culture performance and increase productivity in Escherichia coli (E. coli) fed-batch processes. Designed to deliver a balanced nutrient supply, this hydrolysate-free formulation supports robust cell growth, higher biomass accumulation, and improved product yield across a broad range of E. coli expression strains.

Features of Bacto CD Supreme Feed include:

  • Increased productivity with 10-fold higher plasmid DNA production compared to batch mode
  • Consistency with a 100% chemically defined, hydrolysate-free composition that minimizes variability
  • Streamlined operations using a one-part, ready-to-use formulation that eliminates the need for complex feed preparation
  • Easy integration with Bacto CD Supreme FPM and standard sterilization methods for scalable implementation
  • Simplified compliance with an animal origin–free formulation that supports global regulatory requirements

Developed for compatibility with both Bacto CD Supreme Fermentation Production Medium (FPM) Plus (Cat. Nos. A9378401, A9378501, A9378502) and Bacto CD Supreme FPM (Cat. Nos. A4973701, A4973702), Bacto CD Supreme Feed provides sustained nutrient delivery during fed-batch fermentation that increases biomass accumulation, extends bioreactor runs, and enhances plasmid DNA and protein yield.

As a chemically defined formulation, Bacto CD Supreme Feed eliminates the performance variability associated with peptone-based feeds and supports consistent outcomes across diverse E. coli strains used in plasmid DNA and recombinant protein production.

Offered in convenient liquid and dry powder (DPM) formats, Bacto CD Supreme Feed ensures ease of use, scalability, and reliability from development to large-scale manufacturing.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
Concentración2x
Para utilizar con (aplicación)Microbial Fermentation
FormulaciónAnimal Origin-free, Chemically-defined
Línea de productosBacto CD Supreme
Cantidad10 x 500 mL
Duración de almacenamiento12 Months
FormularioLiquid
Origen de fabricaciónUnited States
Tipo de productoE. coli Feed
EsterilidadSterile
Clase de organismo dianaE. coli
Tipo de pruebaMicrobial Cultivation
Unit SizeCase
Contenido y almacenamiento
Store in the dark at +2°C to +8°C