2-Etilciclohexanona, 99 %, Thermo Scientific Chemicals
2-Etilciclohexanona, 99 %, Thermo Scientific Chemicals
2-Etilciclohexanona, 99 %, Thermo Scientific Chemicals
2-Etilciclohexanona, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Etilciclohexanona, 99 %, Thermo Scientific Chemicals

CAS: 4423-94-3 | C8H14O | 126.199 g/mol
Have Questions?
Número de catálogoCantidad
B20748.14
también denominado B20748-14
25 g
Número de catálogo B20748.14
también denominado B20748-14
Precio (USD)
-
Cantidad:
25 g
Pedido a granel o personalizado
Identificadores químicos
CAS4423-94-3
IUPAC Name2-ethylcyclohexan-1-one
Molecular FormulaC8H14O
InChI KeyWKYYYUWKFPFVEY-UHFFFAOYNA-N
SMILESCCC1CCCCC1=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.4500-1.4530 @ 20°C
2-Ethylcyclohexanone is used as a pharmaceutical intermediate. 2-Methyl and 2-ethyl derivatives of cyclohexanone were allowed to react with deuterium in tBuOD using platinum group metals as catalysts to study the substituent effects in heterogeneous catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
La 2-etilciclohexanona se utiliza como intermediario farmacéutico. Se permitió que los derivados de 2-metilo y 2-etilo de la ciclohexanona reaccionaran con deuterio en tBuOD utilizando metales del grupo del platino como catalizadores para estudiar los efectos sustitutivos en catálisis heterogénea.

Solubilidad
Soluble en alcohol. Insoluble en agua.

Notas
Almacenar a temperatura ambiente. Incompatible con agentes oxidantes fuertes.
RUO – Research Use Only

General References:

  1. Shousuke Teratani, Kazunori Tanaka.; Haruo Ogawa.; Kazuo Taya. Substituent effects in heterogeneous catalysis: VIII. Reactions of 2-methyl- and 2-ethylcyclohexanone with deuterium catalyzed by platinum metals. J. Catal. 1981, 70 (2),347-355 .
  2. Herbert C. Brown.; Milorad M. Rogic.; Michael W. Rathke. Reaction of organoboranes with .alpha.-bromo ketones under the influence of potassium tert-butoxide in tetrahydrofuran. A new technique for the .alpha.-alkylation of ketones.J. Am. Chem. Soc. 1968, 90 (22),6218-6219 .