Thermo Scientific Chemicals

2-Feniletiltrifluoroborato de potasio, 98 %, Thermo Scientific Chemicals

Número de catálogo: H25930.06
5 g, Each
Thermo Scientific Chemicals

2-Feniletiltrifluoroborato de potasio, 98 %, Thermo Scientific Chemicals

Número de catálogo: H25930.06
5 g, Each
Cantidad
Número de catálogo: H25930.06
también denominado H25930-06
Precio (USD)

Identificadores químicos

CAS
329976-74-1
IUPAC Name
potassium trifluoro(2-phenylethyl)boranuide
Molecular Formula
C8H9BF3K
InChI Key
SOZVPVAYKGJPJS-UHFFFAOYSA-N
SMILES
[K+].F[B-](F)(F)CCC1=CC=CC=C1
Appearance (Color)
White
Form
lumpy powder
Purity (DSC)
> 97.5%
Proton NMR
To conform to structure

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text