Thermo Scientific Chemicals

Ácido 3-etil-5-metilisoxazol-4-carboxílico, 97 %, Thermo Scientific Chemicals

Número de catálogo: H27574.09
10 g, Each
Thermo Scientific Chemicals

Ácido 3-etil-5-metilisoxazol-4-carboxílico, 97 %, Thermo Scientific Chemicals

Número de catálogo: H27574.09
10 g, Each
Cantidad
Número de catálogo: H27574.09
también denominado H27574-09
Precio (USD)

Identificadores químicos

CAS
17147-85-2
IUPAC Name
3-ethyl-5-methyl-1,2-oxazole-4-carboxylate
Molecular Formula
C7H8NO3
InChI Key
RSWFHHWVFZWIMV-UHFFFAOYSA-M
SMILES
CCC1=NOC(C)=C1C([O-])=O
Appearance (Color)
White to cream
Form
Powder
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (HPLC)
≥96.0%
Melting Point (clear melt)
118.0-128.0?C

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text