Thermo Scientific Chemicals

Clorhidrato de bencilo de L-serina, 98 %, Thermo Scientific Chemicals

Número de catálogo: H28790.06
5 g, Each
Thermo Scientific Chemicals

Clorhidrato de bencilo de L-serina, 98 %, Thermo Scientific Chemicals

Número de catálogo: H28790.06
5 g, Each
Cantidad
Número de catálogo: H28790.06
también denominado H28790-06
Precio (USD)

Identificadores químicos

CAS
60022-62-0
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Optical Rotation
-11.9 ± 0.4? (C=4 in water)

Descripción

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text