Thermo Scientific Chemicals

Ácido carboxílico 1-Boc-azetidina-3, 97 %, Thermo Scientific Chemicals

Número de catálogo: H28817.06
5 g, Each
Thermo Scientific Chemicals

Ácido carboxílico 1-Boc-azetidina-3, 97 %, Thermo Scientific Chemicals

Número de catálogo: H28817.06
5 g, Each
Cantidad
Número de catálogo: H28817.06
también denominado H28817-06
Precio (USD)

Identificadores químicos

CAS
142253-55-2
IUPAC Name
1-[(tert-butoxy)carbonyl]azetidine-3-carboxylate
Molecular Formula
C9H14NO4
InChI Key
NCADHSLPNSTDMJ-UHFFFAOYSA-M
SMILES
CC(C)(C)OC(=O)N1CC(C1)C([O-])=O
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Melting Point (clear melt)
102.0-108.0?C
Form
Powder
Assay (Silylated GC)
≥96.0%
Appearance (Color)
White

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO - Para uso exclusivo en investigación

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text