Thermo Scientific Chemicals

Ácido 5-fluoropiridin-3-borónico, 98 %, Thermo Scientific Chemicals

Número de catálogo: H52860.03
1 g, Each
Thermo Scientific Chemicals

Ácido 5-fluoropiridin-3-borónico, 98 %, Thermo Scientific Chemicals

Número de catálogo: H52860.03
1 g, Each
Cantidad
Número de catálogo: H52860.03
también denominado H52860-03
Precio (USD)

Identificadores químicos

CAS
872041-86-6
IUPAC Name
(5-fluoropyridin-3-yl)boronic acid
Molecular Formula
C5H5BFNO2
InChI Key
FVEDGBRHTGXPOK-UHFFFAOYSA-N
SMILES
OB(O)C1=CC(F)=CN=C1
Appearance (Color)
White to cream or pale yellow
Assay by NMR
≥94.0%
Form
Crystals or crystalline powder or powder
Proton NMR
Conforms to structure

Descripción

5-Fluoropyridine-3-boronic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
El ácido 5-fluoropiridin-3-borónico se utiliza como intermedio farmacéutico.

Solubilidad
Soluble en metanol caliente.

Notas
Estable en las condiciones de almacenamiento recomendadas. Almacenar lejos de agentes oxidantes.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text