Thermo Scientific Chemicals

N-Alilanilina, 95 %, Thermo Scientific Chemicals

Número de catálogo: H55821.06
5 g, Each
Thermo Scientific Chemicals

N-Alilanilina, 95 %, Thermo Scientific Chemicals

Número de catálogo: H55821.06
5 g, Each
Cantidad
Número de catálogo: H55821.06
también denominado H55821-06
Precio (USD)

Identificadores químicos

CAS
589-09-3
Form
Liquid
Assay (GC)
>94.0%
Refractive Index
1.5620-1.5660 @ 20?C
Appearance (Color)
Clear colorless to orange to brown

Descripción

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
Se utiliza como intermedio farmacéutico.

Solubilidad
Insoluble en agua.

Notas
Mantener alejado de agentes oxidantes y ácidos fuertes.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text