It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
Es una importante materia prima y un intermedio utilizado en la síntesis orgánica, los productos farmacéuticos, los agroquímicos y los colorantes.
Solubilidad
Soluble en agua y 1 % de ácido acético.
Notas
Desecar a – 20 °C. Incompatible con agentes oxidantes.
RUO – Research Use Only
- Nils Drechsler.; Julia Fröbel.; Günther Jahreis.; Mohanraj Gopalswamy.; Jochen Balbach.; Eva Bosse-Doenecke.; Rainer Rudolph. Binding specificity of the ectodomain of the parathyroid hormone receptor. Biophys. Chem. 2011, 154 (2-3),66-72.