Thermo Scientific Chemicals

1-Benzylindole-3-carboxylic acid, 95%, Thermo Scientific Chemicals

Número de catálogo: H66294.06
5 g, Each
Thermo Scientific Chemicals

1-Benzylindole-3-carboxylic acid, 95%, Thermo Scientific Chemicals

Número de catálogo: H66294.06
5 g, Each
Cantidad
Número de catálogo: H66294.06
también denominado H66294-06
Precio (USD)

Identificadores químicos

CAS
27018-76-4
IUPAC Name
1-benzyl-1H-indole-3-carboxylate
Molecular Formula
C16H12NO2
InChI Key
LVYDDRHDOKXFMW-UHFFFAOYSA-M
SMILES
[O-]C(=O)C1=CN(CC2=CC=CC=C2)C2=CC=CC=C12
Appearance (Color)
Pink.
Assay (unspecified)
Water: ≤0.5%
Proton NMR
Conforms
Assay (HPLC)
≥94.0%
Form
Crystalline powder

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text