Lactulosa, 99 %
Lactulosa, 99 %
Lactulosa, 99 %
Thermo Scientific Chemicals

Lactulosa, 99 %

Un azúcar sintético
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
25 g
50 g
100 g
Número de catálogo J60160.18
también denominado J60160-18
Precio (USD)
-
Cantidad:
50 g
Pedido a granel o personalizado
Identificadores químicos
CAS4618-18-2
IUPAC Name(2S,3R,4S,5R,6R)-2-{[(2R,3S,4S,5R)-4,5-dihydroxy-2,5-bis(hydroxymethyl)oxolan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyJCQLYHFGKNRPGE-FCVZTGTOSA-N
SMILESOC[C@H]1O[C@](O)(CO)[C@@H](O)[C@@H]1O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Water Content (Karl Fischer Titration)≤ 2%
Appearance (Color)White
Assay (HPLC)≥ 99%
FormPowder
It is not absorbed in the gut and is frequently used in the treatment of constipation and hepatic encephalopathy. actulose is also a food ingredient, better known as galactofructose, with sweet taste and offering beneficial health benefits on digestive health. Also used as a test of small intestine bacterial overgrowth (SIBO).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubilidad
Soluble en agua (76,4 %) a 30 °C, agua (81 %) a 60 °C y agua (> 86 %) a 90 °C y metanol.

Notas
Almacenar en un lugar fresco. Mantener el recipiente bien cerrado en un lugar seco y bien ventilado. Almacenar lejos de agentes oxidantes.
RUO – Research Use Only

General References:

  1. Tim Vanuytsel et. al. Psychological stress and corticotropin-releasing hormone increase intestinal permeability in humans by a mast cell-dependent mechanism. Gut, 2014, 63(8), 1293-1299.
  2. C P Kelly et. al. Larazotide acetate in patients with coeliac disease undergoing a gluten challenge: a randomised placebo-controlled study. Alimentary Pharmacology and Therapeutics, 2013,37,(2), 252-262.