Thermo Scientific Chemicals

SOC medium, Thermo Scientific Chemicals

Número de catálogo: J60255.AE
100 mL, Each
Thermo Scientific Chemicals

SOC medium, Thermo Scientific Chemicals

Número de catálogo: J60255.AE
100 mL, Each
Cantidad
Número de catálogo: J60255.AE
también denominado J60255-AE
Precio (USD)
Appearance (Color)
Yellow to yellow-brown to brown
Form
Liquid
Assay from Suppliers CofA
Composition specified from vendor COA
Composition
Dextrose/Glucose: 20 mM
Composition
Magnesium Chloride: 10 mM

Descripción

For cultivating recombinant strains of Escherichia coliSOC Medium is used in the final step of bacterial cell transformation to obtain the maximal transformation efficiency of E. coli.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text