Tampón de bloqueo Tween 20, 0,5 % en PBS, 10X
Thermo Scientific Chemicals

Tampón de bloqueo Tween 20, 0,5 % en PBS, 10X

Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 mL
1 L
2 L
Número de catálogo J60304.K2
también denominado J60304-K2
Precio (USD)
-
Cantidad:
1 L
Pedido a granel o personalizado
Especificaciones
Nombre del producto químico o materialTampón de bloqueo
Almacenamiento recomendadoTemperaturas ambientes
Concentración0,5% en PBS (10X)
Forma físicaLíquido
Cantidad1 L
Ver más
Tween 20 blocking buffer solution is used as a washing agent in immunoassays, such as Western blots and ELISAs. It helps to prevent non-specific antibody binding. The buffer is used for washes between each immunoreactions, to remove unbound immunologicals, and eventually for incubation solutions of immunoreagents (labeled antibodies) to reduce unspecific background.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only