Thermo Scientific Chemicals

Ácido 3-hidroxipiridina-2-carboxílico, 98 %, Thermo Scientific Chemicals

Número de catálogo: L07489.03
1 g, Each
Thermo Scientific Chemicals

Ácido 3-hidroxipiridina-2-carboxílico, 98 %, Thermo Scientific Chemicals

Número de catálogo: L07489.03
1 g, Each
Cantidad
Número de catálogo: L07489.03
también denominado L07489-03
Precio (USD)

Identificadores químicos

CAS
874-24-8
IUPAC Name
3-hydroxypyridine-2-carboxylate
Molecular Formula
C6H4NO3
InChI Key
BRARRAHGNDUELT-UHFFFAOYSA-M
SMILES
OC1=CC=CN=C1C([O-])=O
Appearance (Color)
White to dark cream to cream to brown
Assay (Aqueous acid-base Titration)
≥94.0% (non-U.S. specification)
Assay from Suppliers CofA
≥94.0% (U.S. specification)
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥94.0% (non-U.S. specification)

Descripción

3-Hydroxypyridine-2-carboxylic acid is used as a matrix for nucleotides in MALDI mass spectrometry analyses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
El ácido 3-hidroxipiridina-2-carboxílico se utiliza como matriz para nucleótidos en los análisis de espectrometría de masas MALDI.

Solubilidad
Soluble en agua (0,25 g en 10 ml).

Notas
Mantener el recipiente bien cerrado. Almacenar lejos de agentes oxidantes.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text