1-Ciclohexeno-1-carbonitrilo, 98 %, Thermo Scientific Chemicals
1-Ciclohexeno-1-carbonitrilo, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Ciclohexeno-1-carbonitrilo, 98 %, Thermo Scientific Chemicals

Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
2 g
10 g
Número de catálogo L10284.04
también denominado L10284-04
Precio (USD)/ Each
-
Cantidad:
2 g
Pedido a granel o personalizado
Identificadores químicos
CAS1855-63-6
IUPAC Namecyclohex-1-ene-1-carbonitrile
Molecular FormulaC7H9N
InChI KeyGTMWGXABXQTZRJ-UHFFFAOYSA-N
SMILESN#CC1=CCCCC1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4800-1.4840 @ 20?C
Assay (GC)≥97.5%
1-Cyclohexene-1-carbonitrile has been used in the preparation of cyclohexene-1-carboxylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
El 1-ciclohexeno-1-carbonitrilo se ha utilizado en la preparación del ácido ciclohexeno-1- carboxílico.

Solubilidad
No miscible en agua.

Notas
Estable en las condiciones de almacenamiento recomendadas. Incompatible con los agentes oxidantes.
RUO – Research Use Only

General References:

  1. R.L.Cargill; D.F.Bushey; J.J.Good. Alkylation of 1-cyanocyclohexene. J. Org. Chem. 1979, 44, (2),300-301
  2. Kiyoshi Iwai; Henry J.Shine. Ion radicals. 46. Reactions of the adducts of thianthrene and phenoxathiin cation radicals and cyclohexene with nucleophiles. J. Org. Chem. 1981, 46, (2),271-276