Thermo Scientific Chemicals

Clorhidrato de 9-(4-aminofenil)carbazol, 98 %, Thermo Scientific Chemicals

Número de catálogo: L11362.03
1 g, Each
Thermo Scientific Chemicals

Clorhidrato de 9-(4-aminofenil)carbazol, 98 %, Thermo Scientific Chemicals

Número de catálogo: L11362.03
1 g, Each
Cantidad
Número de catálogo: L11362.03
también denominado L11362-03
Precio (USD)

Identificadores químicos

CAS
312700-07-5
IUPAC Name
4-(9H-carbazol-9-yl)aniline hydrochloride
Molecular Formula
C18H15ClN2
InChI Key
CFQROJKMLJRSAL-UHFFFAOYSA-N
SMILES
Cl.NC1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Appearance (Color)
Pale brown or pale blue to purple
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Form
Powder

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text