3,5-dibromo-4-metilanilina, 99 %, Thermo Scientific Chemicals
3,5-dibromo-4-metilanilina, 99 %, Thermo Scientific Chemicals
3,5-dibromo-4-metilanilina, 99 %, Thermo Scientific Chemicals
3,5-dibromo-4-metilanilina, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-dibromo-4-metilanilina, 99 %, Thermo Scientific Chemicals

CAS: 13194-73-5 | C7H7Br2N | 264.948 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Número de catálogo L13101.06
también denominado L13101-06
Precio (USD)
-
Cantidad:
5 g
Pedido a granel o personalizado
Identificadores químicos
CAS13194-73-5
IUPAC Name3,5-dibromo-4-methylaniline
Molecular FormulaC7H7Br2N
InChI KeyAQZDIKCNODUMNY-UHFFFAOYSA-N
SMILESCC1=C(Br)C=C(N)C=C1Br
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
FormCrystals or powder or crystalline powder
Appearance (Color)Pale cream to cream to yellow to brown
Assay (GC)≥98.5%
3,5-Dibromo-4-methylaniline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
La 3,5-dibromo-4-metilanilina se utiliza como producto intermedio farmacéutico.

Solubilidad
No soluble en agua.

Notas
Estables en las condiciones de almacenamiento recomendadas. Incompatible con los agentes oxidantes.
RUO – Research Use Only

General References:

  1. Lisa A.Tobin; Carine Robert; Pratik Nagaria; Saranya Chumsri; William Twaddell; Olga B.Ioffe; George E.Greco; Angela H.Brodie; Alan E.Tomkinson; Feyruz V.Rassool. Targeting abnormal DNA repair in therapy-resistant breast cancers. Mol Cancer Res. 2012, 10,(1), 96-107.
  2. Rene Stangenberg; David Turp; Klaus Mullen. Shape persistent hybrid dendrimers from benzene and triazole via ‘click chemistry’. Tetrahedron. 2014, 70,(19), 3178-3184.