Thermo Scientific Chemicals

2-(1-Piperazinil)pirimidina, 99 %, Thermo Scientific Chemicals

Número de catálogo: L15884.14
25 g, Each
Thermo Scientific Chemicals

2-(1-Piperazinil)pirimidina, 99 %, Thermo Scientific Chemicals

Número de catálogo: L15884.14
25 g, Each
Cantidad
Número de catálogo: L15884.14
también denominado L15884-14
Precio (USD)

Identificadores químicos

CAS
20980-22-7
IUPAC Name
2-(piperazin-1-yl)pyrimidine
Molecular Formula
C8H12N4
InChI Key
MRBFGEHILMYPTF-UHFFFAOYSA-N
SMILES
C1CN(CCN1)C1=NC=CC=N1
Appearance (Color)
Clear
Form
Powder or fused solid or liquid as melt
Transition Temperature
White to yellow or clear colorless to yellow
Assay (GC)
≥98.5%
Refractive Index
1.5865-1.5895 @ 20?C

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text