Thermo Scientific Chemicals

6-cloropiridin-3-metanol, 98 %, Thermo Scientific Chemicals

Número de catálogo: L19285.06
5 g, Each
Thermo Scientific Chemicals

6-cloropiridin-3-metanol, 98 %, Thermo Scientific Chemicals

Número de catálogo: L19285.06
5 g, Each
Cantidad
Número de catálogo: L19285.06
también denominado L19285-06
Precio (USD)

Identificadores químicos

CAS
21543-49-7
IUPAC Name
(6-chloropyridin-3-yl)methanol
Molecular Formula
C6H6ClNO
InChI Key
GOXYBEXWMJZLJB-UHFFFAOYSA-N
SMILES
OCC1=CC=C(Cl)N=C1
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to cream to yellow
Water Content (Karl Fischer Titration)
≤1%

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text