Transfer Assembly Y Manifold
Transfer Assembly Y Manifold
Transfer Assembly Y Manifold
Transfer Assembly Y Manifold
Thermo Scientific™
Transfer Assembly Y Manifold
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. Simplified connectivity and greater process efficiency for most upstream and downstream process operations.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipos de conectorEstilo de los tubos
SH31200.20Conectores AseptiQuik S/Conector Y3/8 in DI de TPE
SH31200.01Conectores AseptiQuik S/Conector Y1/8 in DI de TPE
SH31458.01Conectores AseptiQuik G/Conector Y1/4 in DI de silicona
SH31200.10Conectores AseptiQuik S/Conector Y1/4 in DI de TPE
SH31458.11Conectores AseptiQuik G/Conector Y3/8 in DI de silicona
SH31458.21Conectores AseptiQuik G/Conector Y1/2 in DI de silicona
SH31202.01Conectores AseptiQuik G/Conector Y1/4 in DI de TPE
SH31458.31Conectores AseptiQuik G/Conector Y3/4 in DI de silicona
SH31202.11Conectores AseptiQuik G/Conector Y3/8 in DI de TPE
SH31461.01Conectores AseptiQuik S/Conector Y1/8 in DI de silicona
SH31202.21Conectores AseptiQuik G/Conector Y1/2 in DI de TPE
SH31461.11Conectores AseptiQuik S/Conector Y1/4 in DI de silicona
SH31202.31Conectores AseptiQuik G/Conector Y3/4 in DI de TPE
SH31461.21Conectores AseptiQuik S/Conector Y3/8 in DI de silicona
SH31203.01Conectores AseptiQuik L/Conectores Y3/4 in DI de TPE
SH31203.11Conectores AseptiQuik L/Conector Y1 in DI de TPE
SH31470.01Conectores AseptiQuik L/Conector Y3/4 in DI de silicona
SH31244.01Cuerpo MPX, conector Y, inserto MPX1/2 in DI de TPE
SH31471.01Conectores AseptiQuik L/Conectores Y1 in DI de silicona
SH31246.01Conectores AseptiQuik L/Conectores Y1 in DI de TPE
SH31483.01Cuerpo MPC, conector Y, inserto MPC3/8 in DI de silicona trenzada
SH31310.01Inserto MPC, conector Y, cuerpo MPC3/8 in DI de TPE
SH31485.01Cuerpo MPU, conector Y, inserto MPU3/4 in DI de silicona
SH31311.01Cuerpo MPU, conector Y, inserto MPU3/4 in DI de TPE
SH31486.01Cuerpo MPU, conector Y, inserto MPU1 in DI de silicona trenzada
SH31247.01Inserto MPU, conector Y, cuerpo MPU1 in DI de TPE
SH31487.01Cuerpo MPX, conector Y, inserto MPX1/2 in DI de silicona trenzada
SH31312.01Cuerpo MPU, conector Y, inserto MPUTPE de 1 in
SH31472.01Cuerpo MPC, conector Y, inserto MPC3/8 in DI de silicona
SH31474.01Inserto MPU, conector Y, cuerpo MPU1 in DI de silicona
SH31475.01Inserto MPU, conector Y, cuerpo MPU3/4 in DI de silicona
SH31477.01Cuerpo MPX, conector Y, inserto MPX1/2 in DI de silicona
SH31478.01Conectores AseptiQuik G/Conector Y1/4 in DI de silicona trenzada
SH31478.11Conectores AseptiQuik G/Conector Y3/8 in DI de silicona trenzada
SH31478.21Conectores AseptiQuik G/Conector Y1/2 in DI de silicona trenzada
SH31478.31Conectores AseptiQuik G/Conector Y3/4 in DI de silicona trenzada
SH31479.11Conectores AseptiQuik S/Conector Y1/4 in DI de silicona trenzada
SH31479.21Conectores AseptiQuik S/Conector Y3/8 in DI de silicona trenzada
SH31480.01Conectores AseptiQuik L/Conector Y3/4 in DI de silicona trenzada
SH31481.11Conectores AseptiQuik L/Conector Y1 in DI de silicona trenzada
SH31482.01Conectores AseptiQuik L/Conectores Y1 in DI de silicona trenzada
Número de catálogo SH31200.20
Precio (USD)
-
Tipos de conector:
Conectores AseptiQuik S/Conector Y
Estilo de los tubos:
3/8 in DI de TPE
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
• One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
• Manufactured with BioTitan Retention Device—designed to provide superior universal tubing retention that minimizes the risk of leakage and failure at connection points
• Multiple manufacturing sites ensure security in product supply
• Produced under cGMP conditions in ISO 7 clean rooms to ensure a high level of quality
• Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard BPC products to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common production processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
All Standard Fluid Transfer Assembly products are constructed of capped-end connectors and thermoplastic elastomer (TPE) tubing for use with welders. The Extension Set is available in a variety of tubing sizes and lengths to meet your fluid transfer needs.
These Standard Fluid Transfer Assembly products provide a common set of products that addresses an expansive set of needs. They are manufactured at multiple sites globally to minimize concerns of production interruptions and supply shortages. These products are built to our high quality standards to facilitate the success of your process.
Especificaciones
Tipos de conectorConectores AseptiQuik S/Conector Y
DescripciónTransfer Assembly Y Manifold, 3/8 in. ID TPE, AseptiQuik S connectors and Y connector
Estilo de los tubos3/8 in DI de TPE
Unit Size1 each