Transfer Assembly Jumper Tubing, TPE, Aseptic
Transfer Assembly Jumper Tubing, TPE, Aseptic
Transfer Assembly Jumper Tubing, TPE, Aseptic
Transfer Assembly Jumper Tubing, TPE, Aseptic
Thermo Scientific™
Transfer Assembly Jumper Tubing, TPE, Aseptic
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. Simplified connectivity and greater process efficiency for most upstream and downstream process operations.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipos de conectorEstilo de los tubosTamaño de los tubos
SH31317.03Conectores AseptiQuik G3/4 in DI de TPE15 ft
SH31198.01Conectores AseptiQuik S1/8 in DI de TPE5 ft
SH31198.02Conectores AseptiQuik S1/8 in DI de TPE10 ft
SH31198.03Conectores AseptiQuik S1/8 in DI de TPE15 ft
SH31198.11Conectores AseptiQuik S1/4 in DI de TPE5 ft
SH31198.12Conectores AseptiQuik S1/4 in DI de TPE10 ft
SH31198.13Conectores AseptiQuik S1/4 in DI de TPE15 ft
SH31198.21Conectores AseptiQuik S3/8 in DI de TPE5 ft
SH31198.22Conectores AseptiQuik S3/8 in DI de TPE10 ft
SH31198.23Conectores AseptiQuik S3/8 in DI de TPE15 ft
SH31199.01Conectores AseptiQuik L3/4 in DI de TPE5 ft
SH31199.02Conectores AseptiQuik L3/4 in DI de TPE10 ft
SH31199.03Conectores AseptiQuik L3/4 in DI de TPE15 ft
SH31199.11Conectores AseptiQuik L1 in DI de TPE5 ft
SH31199.12Conectores AseptiQuik L1 in DI de TPE10 ft
SH31199.13Conectores AseptiQuik L1 in DI de TPE15 ft
SH31317.01Conectores AseptiQuik G3/4 in DI de TPE5 ft
SH31317.02Conectores AseptiQuik G3/4 in DI de TPE10 ft
SH31319.01Conectores AseptiQuik G1/4 in DI de TPE5 ft
SH31319.02Conectores AseptiQuik G1/4 in DI de TPE10 ft
SH31319.03Conectores AseptiQuik G1/4 in DI de TPE15 ft
SH31319.11Conectores AseptiQuik G3/8 in DI de TPE5 ft
SH31319.12Conectores AseptiQuik G3/8 in DI de TPE10 ft
SH31319.13Conectores AseptiQuik G3/8 in DI de TPE15 ft
SH31319.21Conectores AseptiQuik G1/2 in DI de TPE5 ft
SH31319.22Conectores AseptiQuik G1/2 in DI de TPE10 ft
SH31319.23Conectores AseptiQuik G1/2 in DI de TPE15 ft
Número de catálogo SH31317.03
Precio (USD)
-
Tipos de conector:
Conectores AseptiQuik G
Estilo de los tubos:
3/4 in DI de TPE
Tamaño de los tubos:
15 ft
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
• One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
• Manufactured with BioTitan Retention Device—designed to provide superior universal tubing retention that minimizes the risk of leakage and failure at connection points
• Multiple manufacturing sites ensure security in product supply
• Produced under cGMP conditions in ISO 7 clean rooms to ensure a high level of quality
• Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard BPC products to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common production processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
All Standard Fluid Transfer Assembly products are constructed of capped-end connectors and thermoplastic elastomer (TPE) tubing for use with welders. The Extension Set is available in a variety of tubing sizes and lengths to meet your fluid transfer needs.
These Standard Fluid Transfer Assembly products provide a common set of products that addresses an expansive set of needs. They are manufactured at multiple sites globally to minimize concerns of production interruptions and supply shortages. These products are built to our high quality standards to facilitate the success of your process.
Especificaciones
Tipos de conectorConectores AseptiQuik G
DescripciónTransfer Assembly Jumper Tubing, 15 ft., 3/4 in. ID TPE, AseptiQuik G connectors
Tamaño de los tubos15 ft
Estilo de los tubos3/4 in DI de TPE
Unit Size1 each