Transfer Assembly Jumper Tubing, Braided Silicone, Aseptic
Transfer Assembly Jumper Tubing, Braided Silicone, Aseptic
Transfer Assembly Jumper Tubing, Braided Silicone, Aseptic
Transfer Assembly Jumper Tubing, Braided Silicone, Aseptic
Thermo Scientific™
Transfer Assembly Jumper Tubing, Braided Silicone, Aseptic
Thermo Scientific™ Standard Fluid Transfer Assemblies with Thermo Scientific™ BioTitan™ Retention Device increases single-use product flexibility. Simplified connectivity and greater process efficiency for most upstream and downstream process operations.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipos de conectorEstilo de los tubosTamaño de los tubos
SH31389.33Conectores AseptiQuik G3/4 in DI de silicona trenzada15 ft
SH31389.01Conectores AseptiQuik G1/4 in DI de silicona trenzada5 ft
SH31389.02Conectores AseptiQuik G1/4 in DI de silicona trenzada10 ft
SH31389.03Conectores AseptiQuik G1/4 in DI de silicona trenzada15 ft
SH31391.11Conectores AseptiQuik S1/4 in DI de silicona trenzada5 ft
SH31391.12Conectores AseptiQuik S1/4 in DI de silicona trenzada10 ft
SH31391.13Conectores AseptiQuik S1/4 in DI de silicona trenzada15 ft
SH31389.11Conectores AseptiQuik G3/8 in DI de silicona trenzada5 ft
SH31389.12Conectores AseptiQuik G3/8 in DI de silicona trenzada10 ft
SH31389.13Conectores AseptiQuik G3/8 in DI de silicona trenzada15 ft
SH31391.21Conectores AseptiQuik S3/8 in DI de silicona trenzada5 ft
SH31391.22Conectores AseptiQuik S3/8 in DI de silicona trenzada10 ft
SH31391.23Conectores AseptiQuik S3/8 in DI de silicona trenzada15 ft
SH31389.21Conectores AseptiQuik G1/2 in DI de silicona trenzada5 ft
SH31389.22Conectores AseptiQuik G1/2 in DI de silicona trenzada10 ft
SH31389.23Conectores AseptiQuik G1/2 in DI de silicona trenzada15 ft
SH31389.31Conectores AseptiQuik G3/4 in DI de silicona trenzada5 ft
SH31389.32Conectores AseptiQuik G3/4 in DI de silicona trenzada10 ft
SH31390.01Conectores AseptiQuik L3/4 in DI de silicona trenzada5 ft
SH31390.02Conectores AseptiQuik L3/4 in DI de silicona trenzada10 ft
SH31390.03Conectores AseptiQuik L3/4 in DI de silicona trenzada15 ft
SH31390.11Conectores AseptiQuik L1 in DI de silicona trenzada5 ft
SH31390.12Conectores AseptiQuik L1 in DI de silicona trenzada10 ft
SH31390.13Conectores AseptiQuik L1 in DI de silicona trenzada15 ft
Número de catálogo SH31389.33
Precio (USD)
-
Tipos de conector:
Conectores AseptiQuik G
Estilo de los tubos:
3/4 in DI de silicona trenzada
Tamaño de los tubos:
15 ft
Thermo Scientific™ Standard Fluid Transfer Assemblies with Thermo Scientific™ BioTitan™ Retention Device increases single-use product flexibility. They connect to our standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
Thermo Scientific™ Standard Fluid Transfer Assemblies with Thermo Scientific™ BioTitan™ Retention Device increases single-use product flexibility. They connect to our standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
• One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
• Manufactured with the BioTitan Retention Device—designed to provide superior universal tubing retention that minimizes the risk of leakage and failure at connection points
• Multiple manufacturing sites ensure security in product supply
• Produced under cGMP conditions in ISO 7 clean rooms to ensure a high level of quality
• Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard BPC products to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common production processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
All Standard Fluid Transfer Assembly products are constructed of capped-end connectors and thermoplastic elastomer (TPE) tubing for use with welders. The Extension Set is available in a variety of tubing sizes and lengths to meet your fluid transfer needs.
These Standard Fluid Transfer Assembly products provide a common set of products that addresses an expansive set of needs. They are manufactured at multiple sites globally to minimize concerns of production interruptions and supply shortages. These products are built to our high quality standards to facilitate the success of your process.
Especificaciones
Tipos de conectorConectores AseptiQuik G
DescripciónTransfer Assembly Jumper Tubing, 15 ft., 3/4 in. ID Braided Silicone, AseptiQuik G Connectors
Para utilizar con (aplicación)Monoclonal antibody production, upstream and downstream applications
CantidadEach
Material del tuboBraided Silicone
Tamaño de los tubos15 ft
Estilo de los tubos3/4 in DI de silicona trenzada
Unit Size1 each
Contenido y almacenamiento
Room Temperature