TRACE™ 1610 High Purity Gas Analyzers
TRACE™ 1610 High Purity Gas Analyzers
TRACE™ 1610 High Purity Gas Analyzers
TRACE™ 1610 High Purity Gas Analyzers
Thermo Scientific™

TRACE™ 1610 High Purity Gas Analyzers

Discover the Thermo Scientific™ TRACE™ 1610 High Purity Gas Analyzers available in dedicated configurations to cover different needs for industrial gas streams analysis.

Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoFunciónCapacidad de columnaTensión
TIGAMS160012Trazas de impurezas en H2, He, Ar, N2 de alta pureza3 capilares120 V
TIGAOX160011Trazas de impurezas en O2 de alta pureza2 capilares120 V
TIGAOX160011-230Trazas de impurezas en O2 de alta pureza2 capilares230 V
TIGAMS160012-230Trazas de impurezas en H2, He, Ar, N2 de alta pureza3 capilares230 V
TIGAHE160013Trazas de impurezas en He de alta purezaPaquete de 2 unidades120 V
TIGAHE160013-230Trazas de impurezas en He de alta purezaPaquete de 2 unidades230 V
IMPC2C316022Trazas de impurezas en hidrocarburos C2 y C3 (ASTM D8098-17, D6159, D5273-18)5 (2 empaquetados, 3 capilares)120 V
IMPC2C316022-230Trazas de impurezas en hidrocarburos C2 y C3 (ASTM D8098-17, D6159, D5273-18)5 (2 empaquetados, 3 capilares)230 V
Número de catálogo TIGAMS160012
Precio (USD)
-
Función:
Trazas de impurezas en H2, He, Ar, N2 de alta pureza
Capacidad de columna:
3 capilares
Tensión:
120 V

Discover our dedicated GC analyzers for low levels of impurities in high purity gases. The Thermo Scientific™ TRACE™ 1610 High Purity Gas analyzers are designed to cover a range of bulk gases such as oxygen, hydrogen, helium, argon, nitrogen, C2 and C3 hydrocarbons.

Rely on the performance and robustness of the variety of Thermo Scientific™ TRACE™ 1610 gas chromatography systems for the analysis of high purity gases, for impurities detection down to ppb-ppt levels.

  • Trace impurities in oxygen: H2, Ar, N2, CO, CO2 and C1-C4
  • Trace impurities in Helium: H2, O2, Ar, N2, CO, CO2 and C1-C4
  • Trace impurities in multiple pure gases (H2, He, Ar, N2): H2, O2, Ar, N2, CO, CO2 and C1-C4
  • Trace impurities in C2 and C3 hydrocarbons: H2, N2, O2, CO, CO2 and C1-C4

Ready to install and run samples

  • Fully configured and factory tested before delivery
  • Full documentation set: start-up and troubleshooting guide, detailed flow and plumbing schematics, spare parts list, method and factory test results

Built to maximize productivity and uptime

  • Spacious independent Auxiliary Oven with easy access to valves, rotors, and actuators
  • Optional third isothermal oven for molecular sieve columns
  • GC features iConnect injector and detector modules removing downtime for troubleshooting and maintenance
  • Heated sample bulkheads ensure complete sample transfer
  • Optimized system footprint to reduce benchspace requirement
Especificaciones
Dimensiones (Largo x Ancho x Alto)67 x 75 x 45 cm (26.4 x 29.5 x 17.7in)
FunciónTrazas de impurezas en H2, He, Ar, N2 de alta pureza
IncluyeGas chromatograph, PDD detector, auxiliary valve oven, three purged valves (one 10-port and two 4-port), sampling loops, columns and method
Tipo de entradaGas Sampling Valve
CantidadEach
Tensión120 V
Capacidad de columna3 capilares
Tipo de detectorPDD
TipoHigh Purity Gas Analyzer
Unit SizeEach