You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
CDMO and Clinical Trial Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Home
Shop All Products
Antibodies
Primary Antibodies
All Primary Antibodies
FMDV-A Monoclonal Antibody
Thermo Scientific™
FMDV-A Monoclonal Antibody
Catalog number:
7500992
Related applications:
Animal Health
Technical Support
Customer Service
Thermo Scientific™
FMDV-A Monoclonal Antibody
Catalog number:
7500992
Related applications:
Animal Health
Technical Support
Customer Service
Catalog Number
7500992
Unit Size
1 mg
Price (BRL)
Contact Us ›
Save to list
Catalog Number
Unit Size
Price (BRL)
7500992
1 mg
Contact Us ›
Save to list
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Monoclonal antibody FMDV-A is directed against serotype FMDV A. The antibody is suited for ELISA and Western blot.
Specifications
Classification
Monoclonal
Host Species
Mouse
Host abrv
Ms
Reactivity
All species
Reactivity abrv
All
Product Line
Prionics™
Figures
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Frequently asked questions (FAQs)
Citations & References
Search citations by name, author, journal title or abstract text
Search