Automation Tips for BioTek™ Pipetting Systems
Actual product may vary
Automation Tips for BioTek™ Pipetting Systems
Automation Tips for BioTek™ Pipetting Systems
Automation Tips for BioTek™ Pipetting Systems
Thermo Scientific™

Automation Tips for BioTek™ Pipetting Systems

Increase pipetting performance with Thermo Scientific™ Automation Tips forBioTek™ Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ColorSterilityFiltered
986-262200 μLClearNon-sterileNon-filtered
985-261100 μLClearSterileNon-filtered
985-262100 μLClearNon-sterileNon-filtered
986-261200 μLClearSterileNon-filtered
Catalog number 986-262
Price (BRL)
-
Volume (Metric):
200 μL
Color:
Clear
Sterility:
Non-sterile
Filtered:
Non-filtered
Increase pipetting performance with Thermo Scientific™ Automation Tips forBioTek™ Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: BioTek Precision and Precision XS.
Features:
  • Available as sterile and filtered.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Specifications
ColorClear
FilteredNon-filtered
For Use With (Equipment)BioTek™ Pipetting System
FormatRack
Product LineBioTek
SterilityNon-sterile
Tip StyleLow Retention
Volume (Metric)200 μL
Unit SizeEach