Thermo Scientific™
Smart-Vue™ Client Server Software
Catalog number: SV602-500-LSB
Thermo Scientific™
Smart-Vue™ Client Server Software
Catalog number: SV602-500-LSB
Automate monitoring of vital laboratory equipment parameters with Thermo Scientific™ Smart-Vue™ Client Server Software. Find peace of mind knowing your work is fully monitored 24/7 with immediate notifications reaching you even when you are away from the lab. This software provides a plug and play solution with an intuitive interface.
Catalog Number
SV602-500-LSB
Unit Size
Each
Description
OPC Server for Smart-Vue
Price (BRL)
Full specifications
TypeOPC Server
DescriptionOPC Server for Smart-Vue
IncludesDriver for Smart-Vue server, user manuals, installation guide
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Equipment)Smart-Vue™ Software
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Smart-Vue Client Server Software (v 1.0) and User Manuals
Price (BRL)
Full specifications
TypeUser Manual Software
DescriptionSmart-Vue Client Server Software (v 1.0) and User Manuals
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
ModelSV600-500-LSB
For Use With (Equipment)Refrigerator, Freezer, LN2 Storage, CO2 Incubator, Oven
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Smart-Vue Client User Manuals, Quick Installation Guides, and Software Drivers for all system receivers and repeaters
Price (BRL)
Full specifications
TypeUser Manuals, Quick Installation Guide, Drivers
DescriptionSmart-Vue Client User Manuals, Quick Installation Guides, and Software Drivers for all system receivers and repeaters
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Equipment)Refrigerator, Freezer, LN2 Storage, CO2 Incubator, Oven
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Smart-Vue Software 2.0, 20 sensors
Price (BRL)
Full specifications
TypeSensor Software
DescriptionSmart-Vue Software 2.0, 20 sensors
IncludesSoftware drivers for all system receivers and repeaters; includes user manuals / quick installation guides.
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Equipment)Refrigerator, Freezer, LN2 Storage, CO2 Incubator, Oven
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Smart-Vue Software 2.0, 49 sensors
Price (BRL)
Full specifications
TypeSensor Software
DescriptionSmart-Vue Software 2.0, 49 sensors
IncludesSoftware drivers for all system receivers and repeaters; includes user manuals / quick installation guides.
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Equipment)Refrigerator, Freezer, LN2 Storage, CO2 Incubator, Oven
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Smart-Vue Software 2.0, 99 sensors
Price (BRL)
Full specifications
TypeSensor Software
DescriptionSmart-Vue Software 2.0, 99 sensors
IncludesSoftware drivers for all system receivers and repeaters; includes user manuals / quick installation guides.
For Use With (Equipment)Refrigerator, Freezer, LN2 Storage, CO2 Incubator, Oven
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Smart-Vue Software 2.0, unlimited sensors
Price (BRL)
Full specifications
TypeSensor Software
DescriptionSmart-Vue Software 2.0, unlimited sensors
IncludesSoftware drivers for all system receivers and repeaters; includes user manuals / quick installation guides.
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Equipment)Refrigerator, Freezer, LN2 Storage, CO2 Incubator, Oven
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Smart-Vue Software 2.0, 20-49 sensors
Price (BRL)
Full specifications
TypeSensor Software
DescriptionSmart-Vue Software 2.0, 20-49 sensors
IncludesSoftware drivers for all system receivers and repeaters; includes user manuals / quick installation guides.
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Equipment)Refrigerator, Freezer, LN2 Storage, CO2 Incubator, Oven
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Smart-Vue Software 2.0, 20-99 sensors
Price (BRL)
Full specifications
TypeSensor Software
DescriptionSmart-Vue Software 2.0, 20-99 sensors
IncludesSoftware drivers for all system receivers and repeaters; includes user manuals / quick installation guides.
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Equipment)Refrigerator, Freezer, LN2 Storage, CO2 Incubator, Oven
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Smart-Vue Software 2.0, 20-unlimited sensors
Price (BRL)
Full specifications
TypeSensor Software
DescriptionSmart-Vue Software 2.0, 20-unlimited sensors
IncludesSoftware drivers for all system receivers and repeaters; includes user manuals / quick installation guides.
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Equipment)Refrigerator, Freezer, LN2 Storage, CO2 Incubator, Oven
Unit SizeEach
Showing 10 of 17Show more
Catalog NumberSpecificationsUnit SizeDescriptionPrice (BRL)
SV602-500-LSBFull specifications
EachOPC Server for Smart-VueRequest A Quote
TypeOPC Server
DescriptionOPC Server for Smart-Vue
IncludesDriver for Smart-Vue server, user manuals, installation guide
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Equipment)Smart-Vue™ Software
Unit SizeEach
SV600-500-LSBFull specifications
EachSmart-Vue Client Server Software (v 1.0) and User ManualsRequest A Quote
TypeUser Manual Software
DescriptionSmart-Vue Client Server Software (v 1.0) and User Manuals
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
ModelSV600-500-LSB
For Use With (Equipment)Refrigerator, Freezer, LN2 Storage, CO2 Incubator, Oven
Unit SizeEach
SV601-500-LSBFull specifications
EachSmart-Vue Client User Manuals, Quick Installation Guides, and Software Drivers for all system receivers and repeatersRequest A Quote
TypeUser Manuals, Quick Installation Guide, Drivers
DescriptionSmart-Vue Client User Manuals, Quick Installation Guides, and Software Drivers for all system receivers and repeaters
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Equipment)Refrigerator, Freezer, LN2 Storage, CO2 Incubator, Oven
Unit SizeEach
SV603-500-LSBFull specifications
EachSmart-Vue Software 2.0, 20 sensorsRequest A Quote
TypeSensor Software
DescriptionSmart-Vue Software 2.0, 20 sensors
IncludesSoftware drivers for all system receivers and repeaters; includes user manuals / quick installation guides.
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Equipment)Refrigerator, Freezer, LN2 Storage, CO2 Incubator, Oven
Unit SizeEach
SV604-500-LSBFull specifications
EachSmart-Vue Software 2.0, 49 sensorsRequest A Quote
TypeSensor Software
DescriptionSmart-Vue Software 2.0, 49 sensors
IncludesSoftware drivers for all system receivers and repeaters; includes user manuals / quick installation guides.
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Equipment)Refrigerator, Freezer, LN2 Storage, CO2 Incubator, Oven
Unit SizeEach
SV605-500-LSBFull specifications
EachSmart-Vue Software 2.0, 99 sensorsRequest A Quote
TypeSensor Software
DescriptionSmart-Vue Software 2.0, 99 sensors
IncludesSoftware drivers for all system receivers and repeaters; includes user manuals / quick installation guides.
For Use With (Equipment)Refrigerator, Freezer, LN2 Storage, CO2 Incubator, Oven
Unit SizeEach
SV606-500-LSBFull specifications
EachSmart-Vue Software 2.0, unlimited sensorsRequest A Quote
TypeSensor Software
DescriptionSmart-Vue Software 2.0, unlimited sensors
IncludesSoftware drivers for all system receivers and repeaters; includes user manuals / quick installation guides.
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Equipment)Refrigerator, Freezer, LN2 Storage, CO2 Incubator, Oven
Unit SizeEach
SV607-500-LSBFull specifications
EachSmart-Vue Software 2.0, 20-49 sensorsRequest A Quote
TypeSensor Software
DescriptionSmart-Vue Software 2.0, 20-49 sensors
IncludesSoftware drivers for all system receivers and repeaters; includes user manuals / quick installation guides.
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Equipment)Refrigerator, Freezer, LN2 Storage, CO2 Incubator, Oven
Unit SizeEach
SV608-500-LSBFull specifications
EachSmart-Vue Software 2.0, 20-99 sensorsRequest A Quote
TypeSensor Software
DescriptionSmart-Vue Software 2.0, 20-99 sensors
IncludesSoftware drivers for all system receivers and repeaters; includes user manuals / quick installation guides.
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Equipment)Refrigerator, Freezer, LN2 Storage, CO2 Incubator, Oven
Unit SizeEach
SV609-500-LSBFull specifications
EachSmart-Vue Software 2.0, 20-unlimited sensorsRequest A Quote
TypeSensor Software
DescriptionSmart-Vue Software 2.0, 20-unlimited sensors
IncludesSoftware drivers for all system receivers and repeaters; includes user manuals / quick installation guides.
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Equipment)Refrigerator, Freezer, LN2 Storage, CO2 Incubator, Oven
Unit SizeEach
Showing 10 of 17Show more
Intelligent Plug & Play Simplicity
  • Installation Wizard for simplified, step-by-step process
  • Service Discovery Protocol (SDP) enables every module to send signal to receiver—identifies most efficient communication route based on RF signal strength
  • Module automatically establishes new path to receiver if no communication during 6-hour period
  • Back-up system with memory capacity up to 3000 points, with uninterruptible power for continuous data logging
Intuitive Dashboard
  • Daily usage is simplified
  • Single-click export of reports to Microsoft™ Excel™, Word™ or Adobe™ PDF formats
  • Flexible creation of graphs summarizing data from individual or multiple sensors
  • Visual depiction of RF signal strength an QoS as monitored by each module
Data Traceability & Security
  • Secure, customized levels of user access
  • Audit trails
  • Archived alarms
  • Password protection
  • Secure digital transmission and data back-ups
Numerous Configuration Capabilities
  • Combines RF and TCP/IP network technologies
  • Share equipment data over entire network instantly
  • Download simple plug-in to visualize equipment data readings from any workstation
Compatible with:
Refrigerators, Freezers, LN2 Storage, CO2 Incubators, Clean Rooms, Storage Areas, Ovens
Recommended for:
Ideal for use in highly controlled environments, including pharmaceutical production, biotechnology, IVF clinics, clinical studies and diagnostic laboratories.
Availability of Smart-Vue products vary by country. Please ask your local Sales Representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements. Specifications, terms and pricing are subject to change. Not all products are available in all countries.