Search
| Número do catálogo | For Use With (Equipment) | Sample Capacity |
|---|---|---|
| 6823.0018 | Dionex UltiMate 3000 Autosampler WPS-3000 TBPL Analytical | 20 μL |
| 6821.0032 | Dionex UltiMate 3000 Autosampler WPS-3000 TBPL Analytical | 125 μL |
| 6823.0016 | Dionex UltiMate 3000 Autosampler WPS-3000 TBPL Analytical | 5 μL |
| 6823.0017 | Dionex UltiMate 3000 Autosampler WPS-3000 TBPL Analytical | 10 μL |
| 6823.0019 | Dionex UltiMate 3000 Autosampler WPS-3000 TBPL Analytical, Dionex UltiMate 3000 Autosampler WPS-3000 TBFC Analytical | 50 μL |
| 6823.0020 | Dionex UltiMate 3000 Autosampler WPS-3000 TBPL Analytical, Dionex UltiMate 3000 Autosampler WPS-3000 TBFC Analytical | 250 μL |
Order the Thermo Scientific™ Biocompatible Sample Loops (5-250μL) you need for Thermo Scientific™ Dionex™ UltiMate™ WPS-3000TBPL pulled loop autosamplers.