Product Image
Thermo Scientific™

MP35N Biocompatible Sample Loops for Split-Loop Autosamplers

Order the MP35N biocompatible sample loop (25-500μL) you need for use in Thermo Scientific™ Dionex™ UltiMate™ split-loop autosamplers.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoSample Capacity
6841.2453250 μL
6841.2451100 μL
6841.245225 μL
6841.2454500 μL
Número do catálogo 6841.2453
Preço (BRL)
-
Sample Capacity:
250 μL
Order the Thermo Scientific™ MP35N Biocompatible Sample Loop you need for use in Thermo Scientific™ Dionex™ UltiMate™ split-loop autosamplers. Available in sizes from 25 to 500μL and made of a nickel alloy, the sample loops incorporate Thermo Scientific™ Dionex™ Viper™ Fingertight Fittings which eliminate dead volumes and ensure easy, perfect connections in your HPLC or UHPLC system every time.
Especificações
Biocompatible VersionYes
For Use With (Equipment)Dionex UltiMate 3000 Split-Loop Autosampler
MaterialMP35N
Sample Capacity250 μL
TypeSample Loop
Unit SizeEach