Specifications
DescriptionEpoxy coated
Dimensions (L x W x H)3.4 x 5 x 8.5 in. (8.63 x 12.7 x 21.59 cm)
For Use With (Application)Perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use
For Use With (Equipment)01-828A, 01-828-031, 01-830A, 01-826-1, 01-827A, 01-829A, 01-815 11 H x 8.5 in. W Biohazard Bags biohazard bag, Clavies™ autoclavable bags (01-815-6)
Height (English)8.5 in.
Height (Metric)21.59 cm
IncludesRectangular Stainless-Steel Wire Frame Only
Length (Metric)8.63 cm
MountingBenchtop
Width (Metric)12.7 cm
AutoclavableAutoclavable
ColorOrange
DisposableSingle-use
MaterialSteel Wire
Product TypeBiohazard Bag Holder
Width (English)5 in.
Unit SizeEach