Nunc™ FluoroNunc™/LumiNunc™ 96-Well Plates
Actual product may vary
Thermo Scientific™

Nunc™ FluoroNunc™/LumiNunc™ 96-Well Plates

Use these well plates for quantitative and qualitative solid phase immunoassays (e.g., FLISA) and binding assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
437958Clear
437796White
Catalog number 437958
Price (CNY)
-
Color:
Clear

Use Thermo Scientific™ Nunc FluoroNunc/LumiNunc 96-Well Solid plates for quantitative and qualitative solid phase immunoassays (e.g., FLISA) and binding assays. Thermo Scientific Nunc Immuno plates offer lot-to-lot reliability, well-to-well reproducibility, and are backed by industry-leading quality standards and certifications.

  • Plate has SBS footprint—fits standard equipment
  • Available with flat-bottom or C-shaped wells
  • Choice of PolySorp™ or MaxiSorp™ surface
  • Offered as black, white or clear plates
  • Certified binding homogeneity/reproducibility

Choice of Surface

  • PolySorp - Hydrophobic surface that preferentially binds biomolecules that have hydrophobic domains

  • MaxiSorp - Hydrophilic surface that is optimized for binding IgG and also displays increased binding of many other proteins and biomolecules that possess hydrophilic/hydrophobic character. Available with or without lot specific

  • performance testing via an IgG binding assay (Well to Well %CV less than 5% for clear plates and less than 10% for colored plates). Tested items come with a certificate

White

  • Gives maximum reflection
  • Minimum autofluorescence and autoluminescence

Black

  • Minimizes background in fluorescence reading
  • Minimizes back-scattered light
  • Minimizes crosstalk

Clear

  • For time-resolved fluorescence

Compatible with:

Compatible with lids for MicroWell™ plates, sealing tape and breathable membrane

Specifications
Binding PropertyHydrophilic
Binding TypeAdsorption
Certifications/ComplianceCertified for the absorption of IgG; well to well %CV of less than 5%.
Culture Area2.5 cm2
Detection MethodColorimetric
LidWith Lid
No. per Case60
No. per Pack5
Packaging60 Cases, 5 Pk.
Plate BlockingNone
Product TypeMicroplate
Target MoleculeAntibodies (Glycoproteins)
Volume (Metric) Working250 μL
Array8 x 12
ColorClear
FormatStandard
MaterialPolystyrene
No. of Wells96
Product LineNunc
Surface TreatmentMaxiSorp
Well ShapeCurved Edges, Flat Bottom (C-well)
Unit SizeCase of 60