Niobium foil, 0.127mm (0.005in) thick, 99.97% (metals basis excluding Ta), Ta ∣<0.06%
Niobium foil, 0.127mm (0.005in) thick, 99.97% (metals basis excluding Ta), Ta ∣<0.06%
Niobium foil, 0.127mm (0.005in) thick, 99.97% (metals basis excluding Ta), Ta ∣<0.06%
Thermo Scientific Chemicals

Niobium foil, 0.127mm (0.005in) thick, 99.97% (metals basis excluding Ta), Ta ∣<0.06%

CAS: 7440-03-1 | Nb | 92.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
25 x 100 mm
50 x 100 mm
Catalog number 000236.FQ
also known as 000236-FQ
Price (EUR)
-
Quantity:
25 x 100 mm
Request bulk or custom format
Chemical Identifiers
CAS7440-03-1
IUPAC Nameniobium
Molecular FormulaNb
InChI KeyGUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 0.127 mm (0.005 in.) thick
Impurity contentTantalum: ≤0.06%
Total Metal Impurities≤0.03% (excluding Ta)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only