Selenium shot, amorphous, 1-6mm (0.04-0.24in), Puratronic, 99.999+% (metals basis)
Selenium shot, amorphous, 1-6mm (0.04-0.24in), Puratronic, 99.999+% (metals basis)
Selenium shot, amorphous, 1-6mm (0.04-0.24in), Puratronic, 99.999+% (metals basis)
Thermo Scientific Chemicals

Selenium shot, amorphous, 1-6mm (0.04-0.24in), Puratronic, 99.999+% (metals basis)

CAS: 7782-49-2 | Se | 78.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 000290.30
also known as 000290-30
Price (EUR)
311,00
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
311,00
Each
Specifications
AppearanceVarious colors shot
Assay Percent Range>99.999% (Metals basis)
CAS7782-49-2
ChEBICHEBI:27568
Chemical Name or MaterialSelenium
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only