Tantalum wire, 0.127mm (0.005in) dia, 99.9% (metals basis)
Tantalum wire, 0.127mm (0.005in) dia, 99.9% (metals basis)
Tantalum wire, 0.127mm (0.005in) dia, 99.9% (metals basis)
Thermo Scientific Chemicals

Tantalum wire, 0.127mm (0.005in) dia, 99.9% (metals basis)

CAS: 7440-25-7 | Ta | 180.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
5 m
Catalog number 000977.G1
also known as 000977-G1
Price (EUR)
-
Quantity:
1 m
Request bulk or custom format
Chemical Identifiers
CAS7440-25-7
IUPAC Nametantalum
Molecular FormulaTa
InChI KeyGUVRBAGPIYLISA-UHFFFAOYSA-N
SMILES[Ta]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.9% (metals basis)
FormWire, 0.127mm (0.005in) diameter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only