Carbonylhydridotris(triphenylphosphine)rhodium(I), Rh 10.0% min
Carbonylhydridotris(triphenylphosphine)rhodium(I), Rh 10.0% min
Carbonylhydridotris(triphenylphosphine)rhodium(I), Rh 10.0% min
Thermo Scientific Chemicals

Carbonylhydridotris(triphenylphosphine)rhodium(I), Rh 10.0% min

Hydrogenation, hydrosilation, isomerization, carbonylation, hydroformylation, oxidation | CAS: 17185-29-4 | C55H46OP3Rh | 918.80 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.1 g
1 g
5 g
Catalog number 010016.03
also known as 010016-03
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS17185-29-4
IUPAC Namemethanidylidyneoxidanium tris(triphenylphosphane) λ¹-rhodium
Molecular FormulaC55H46OP3Rh
InChI KeyAACIZACVKFEETJ-UHFFFAOYSA-N
SMILES[RhH].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Rh 10.0% min
Hydrogenation, hydrosilation, isomerization, carbonylation, hydroformylation, oxidation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only