Molybdenum plate, 2.5mm (0.098in) thick, 99.95% (metals basis)
Molybdenum plate, 2.5mm (0.098in) thick, 99.95% (metals basis)
Molybdenum plate, 2.5mm (0.098in) thick, 99.95% (metals basis)
Thermo Scientific Chemicals

Molybdenum plate, 2.5mm (0.098in) thick, 99.95% (metals basis)

CAS: 7439-98-7 | Mo | 95.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
Catalog number 010047.FI
also known as 010047-FI
Price (EUR)
-
Quantity:
50 x 50 mm
Request bulk or custom format
Chemical Identifiers
CAS7439-98-7
IUPAC Namemolybdenum
Molecular FormulaMo
InChI KeyZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES[Mo]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only