Copper shot, 1-10mm (0.04-0.4in), 99.9% (metals basis)
Copper shot, 1-10mm (0.04-0.4in), 99.9% (metals basis)
Copper shot, 1-10mm (0.04-0.4in), 99.9% (metals basis)
Thermo Scientific Chemicals

Copper shot, 1-10mm (0.04-0.4in), 99.9% (metals basis)

CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 010159.36
also known as 010159-36
Price (EUR)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red
FormShot
Total Metal Impurities≤ 0.1%
Particle Size1-10mm
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only