Nickel wire, 0.5mm (0.02in) dia, annealed, 99.5% (metals basis)
Nickel wire, 0.5mm (0.02in) dia, annealed, 99.5% (metals basis)
Nickel wire, 0.5mm (0.02in) dia, annealed, 99.5% (metals basis)
Thermo Scientific Chemicals

Nickel wire, 0.5mm (0.02in) dia, annealed, 99.5% (metals basis)

Form: Approx.1.75g/m, Temper: soft | CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 m
25 m
100 m
500 m
Catalog number 010250.G9
also known as 010250-G9
Price (EUR)
97,65
Online exclusive
108,00
Save 10,35 (10%)
Each
Quantity:
25 m
Request bulk or custom format
Price (EUR)
97,65
Online exclusive
108,00
Save 10,35 (10%)
Each
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire
Transition TemperatureSilver to gray
Diameter0.45 - 0.55mm
Total Metal Impurities0.5% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only