Ammonium hydrogen phosphate, tech., P{2}O{5} 53% min
Ammonium hydrogen phosphate, tech., P{2}O{5} 53% min
Ammonium hydrogen phosphate, tech., P{2}O{5} 53% min
Thermo Scientific Chemicals

Ammonium hydrogen phosphate, tech., P{2}O{5} 53% min

Corrosion inhibitors, in fermentation, as flux for soldering tin, copper, brass, and zinc | CAS: 7783-28-0 | H9N2O4P | 132.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 010301.A1
also known as 010301-A1
Price (EUR)
54,80
Each
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
54,80
Each
Chemical Identifiers
CAS7783-28-0
IUPAC Namephosphoric acid diamine
Molecular FormulaH9N2O4P
InChI KeyMNNHAPBLZZVQHP-UHFFFAOYSA-N
SMILESN.N.OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentSodium, Na: ≤0.005% (≤50ppm)
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay from Supplier's CofA≥98.0%
Chloride content≤0.001% (≤10ppm)
View more
Ammonium hydrogen phosphate is used as a fertilizer, as a fire retardant, as a yeast nutrient in winemaking and brewing mead, as an additive in some brands of cigarettes, in purifying sugar and to control precipitation of alkali-soluble and acid-insoluble colloidal dyes on wool.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only