Sodium ingot, 99.8% (metals basis)
Sodium ingot, 99.8% (metals basis)
Sodium ingot, 99.8% (metals basis)
Sodium ingot, 99.8% (metals basis)
Thermo Scientific Chemicals

Sodium ingot, 99.8% (metals basis)

CAS: 7440-23-5 | Na | 22.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
450 g
5 x 450 g
Catalog number 010342.35
also known as 010342-35
Price (EUR)
-
Quantity:
450 g
Request bulk or custom format
Chemical Identifiers
CAS7440-23-5
IUPAC Namesodium
Molecular FormulaNa
InChI KeyKEAYESYHFKHZAL-UHFFFAOYSA-N
SMILES[Na]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey
FormIngot
Total Metal Impurities0.2% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

How is Sodium ingot, 99.8% (metals basis) packaged?

Sodium ingot, 99.8% (metals basis) is packaged dry (not in oil) in a plastic bag, flushed with argon, and placed in a metal can.