Titanium cube, 9.5x9.5x9.5mm (0.37x0.37x0.37in), 99.99% (metals basis)
Titanium cube, 9.5x9.5x9.5mm (0.37x0.37x0.37in), 99.99% (metals basis)
Titanium cube, 9.5x9.5x9.5mm (0.37x0.37x0.37in), 99.99% (metals basis)
Thermo Scientific Chemicals

Titanium cube, 9.5x9.5x9.5mm (0.37x0.37x0.37in), 99.99% (metals basis)

CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 Pc.
5 Pc.
Catalog number 010380.KG
also known as 010380-KG
Price (EUR)
-
Quantity:
1 Pc.
Request bulk or custom format
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
FormCube, 9.5mm (0.37in) square.
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.99% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only