Tungsten wire, 1.0mm (0.04in) dia, 99.95% (metals basis)
Tungsten wire, 1.0mm (0.04in) dia, 99.95% (metals basis)
Tungsten wire, 1.0mm (0.04in) dia, 99.95% (metals basis)
Thermo Scientific Chemicals

Tungsten wire, 1.0mm (0.04in) dia, 99.95% (metals basis)

CAS: 7440-33-7 | W | 183.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
5 m
25 m
Catalog number 010411.G1
also known as 010411-G1
Price (EUR)
-
Quantity:
1 m
Request bulk or custom format
Chemical Identifiers
CAS7440-33-7
IUPAC Nametungsten
Molecular FormulaW
InChI KeyWFKWXMTUELFFGS-UHFFFAOYSA-N
SMILES[W]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 1.0mm (0.04in) diameter
Total Metal Impurities≤0.05%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only