Zinc shot, 10mm (0.4in) dia x 2mm (0.08in) thick, 99.99% (metals basis)
Zinc shot, 10mm (0.4in) dia x 2mm (0.08in) thick, 99.99% (metals basis)
Zinc shot, 10mm (0.4in) dia x 2mm (0.08in) thick, 99.99% (metals basis)
Thermo Scientific Chemicals

Zinc shot, 10mm (0.4in) dia x 2mm (0.08in) thick, 99.99% (metals basis)

CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 010440.22
also known as 010440-22
Price (EUR)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS7440-66-6
IUPAC Namezinc
Molecular FormulaZn
InChI KeyHCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤ 0.01%
Appearance (Color)Gray
Diameter8-10mm
FormShot
Thickness1.8-2.2mm
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only