Palladium(II) 2,4-pentanedionate, Pd 34.7%
Palladium(II) 2,4-pentanedionate, Pd 34.7%
Palladium(II) 2,4-pentanedionate, Pd 34.7%
Thermo Scientific Chemicals

Palladium(II) 2,4-pentanedionate, Pd 34.7%

Carbonylation, oligomerization | CAS: 14024-61-4 | C10H14O4Pd | 304.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 010517.03
also known as 010517-03
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS14024-61-4
IUPAC Namepalladium(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14O4Pd
InChI KeyJKDRQYIYVJVOPF-FDGPNNRMSA-L
SMILES[Pd++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to orange
FormPowder
Assay (unspecified)Palladium (Pd): 33.9-35.3%
It is used in carbonylation, oligomerization, and as a catalyst precursor for various synthetic organic transformations, for example in the preparation of complexes to efficiently catalyze Heck reaction. Useful in the decarboxylative cross-coupling of arylcarboxylic acids with aryl halides, in the preparation of monodisperse nanoparticles of CuPd alloy and in several cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only