Tetrakis(triphenylphosphine)palladium(0), 99.8% (metals basis), Pd 9% min
Tetrakis(triphenylphosphine)palladium(0), 99.8% (metals basis), Pd 9% min
Tetrakis(triphenylphosphine)palladium(0), 99.8% (metals basis), Pd 9% min
Thermo Scientific Chemicals

Tetrakis(triphenylphosphine)palladium(0), 99.8% (metals basis), Pd 9% min

Hydrosilation, isomerization, carbonylation | CAS: 14221-01-3 | C72H60P4Pd | 1155.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
10 g
Catalog number 010548.09
also known as 010548-09
Price (EUR)
473,00
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
473,00
Each
Chemical Identifiers
CAS14221-01-3
IUPAC Nametetrakis(triphenylphosphane) palladium
Molecular FormulaC72H60P4Pd
InChI KeyNFHFRUOZVGFOOS-UHFFFAOYSA-N
SMILES[Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Pd 9% min
Total Metal Impurities0.2% max.
Appearance (Color)Yellow to yellow/green
FormPowder
Widely used as a catalyst for palladium-catalyzed coupling reactions including Suzuki coupling, Sonogashira coupling, Heck reaction, Negishi coupling and Stille coupling. Used in isomerization, carbonylation, oxidation, and C-C bond formation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only