Aluminum nitrate hydrate, Puratronic™, 99.999% (metals basis excluding Hg)
Aluminum nitrate hydrate, Puratronic™, 99.999% (metals basis excluding Hg)
Aluminum nitrate hydrate, Puratronic™, 99.999% (metals basis excluding Hg)
Thermo Scientific Chemicals

Aluminum nitrate hydrate, Puratronic™, 99.999% (metals basis excluding Hg)

CAS: 25838-59-9 | AlN3O9 | 212.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number 010626.06
also known as 010626-06
Price (EUR)
38,40
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
38,40
Each
Chemical Identifiers
CAS25838-59-9
IUPAC Namealuminium(3+) trinitrate
Molecular FormulaAlN3O9
InChI KeyJLDSOYXADOWAKB-UHFFFAOYSA-N
SMILES[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Aluminum nitrate hydrate is used to produce alumina for the preparation of insulating papers, and in the extraction of actinide elements. It is also used in the preparation of rare earth aluminates, in the heating elements of cathode ray tubes, as a precursor for sol-gel synthesis of alumina, and on transformer core laminates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only