Antimony(III) oxide, Puratronic™, 99.999% (metals basis)
Antimony(III) oxide, Puratronic™, 99.999% (metals basis)
Antimony(III) oxide, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Antimony(III) oxide, Puratronic™, 99.999% (metals basis)

CAS: 1309-64-4 | O3Sb2 | 291.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
50 g
100 g
Catalog number 010641.14
also known as 010641-14
Price (EUR)
175,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
175,00
Each
Chemical Identifiers
CAS1309-64-4
IUPAC Namediantimony(3+) trioxidandiide
Molecular FormulaO3Sb2
InChI KeyGHPGOEFPKIHBNM-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Sb+3].[Sb+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (special)+/- 2.5 % of Theoretical value (Sb)
Total Metal Impurities≤0.001% (10 ppm)
Antimony(III) oxide finds use as flame retardant wherein it synergistically acts with halogenated materials. It is employed in opacifying glasses, ceramics and enamels. In the preparation of optical and ruby glass, antimony trioxide serves as a bubble remover. Further, it is a useful catalyst in the production of polyethylene terephthalate (PET plastic) and the vulcanization of rubber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only