Bismuth(III) nitrate hydrate, Puratronic™, 99.999% (metals basis)
Bismuth(III) nitrate hydrate, Puratronic™, 99.999% (metals basis)
Bismuth(III) nitrate hydrate, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Bismuth(III) nitrate hydrate, Puratronic™, 99.999% (metals basis)

CAS: 10361-44-1 | BiN3O9 | 394.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 010657.22
also known as 010657-22
Price (EUR)
334,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
334,00
Each
Chemical Identifiers
CAS10361-44-1
IUPAC Namebismuth(3+) trinitrate
Molecular FormulaBiN3O9
InChI KeyPPNKDDZCLDMRHS-UHFFFAOYSA-N
SMILES[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
It is used in electronic ceramics and fluorescent paints. It is employed as an oxidant for selective oxidation of sulfides to sulfoxides, as a nitrating reagent for synthesizing nitro aromatic compounds, and as a precursor for Dragendorff reagent for use in TLC stain and for testing the presence of alkaloids. On silica gel support, it has been reported to be an efficient catalyst for the Biginelli reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only